Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (+/-)-Atenolol

Catalog No. 5054160001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-541-60001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-541-60001 Supplier MilliporeSigma™ Supplier No. 5054160001
Add to Cart
Add to Cart

Selective β1 adrenergic blocking ag

Selective β1 adrenergic blocking agent. An anti-hypertensive and anti-arrhythmic agent. Reported to diminish cerebrospinal fluid production.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins β1 adrenergic receptor
Molecular Formula C14H22N2O3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.