Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ATIC Dimerization Inhibitor

Catalog No. 11849010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-849-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-849-010MG Supplier MilliporeSigma™ Supplier No. 11849010MG
Add to Cart
Add to Cart

A cell-permeable dipeptide that reversibly blocks the homodimerization of aminoimidazole carboxamide ribonucleotide transformylase/inosine nomophosphate cyclohydrolase (ATIC) and inhibits its activity (Ki = 691 nM, Kd = 2

A cell-permeable dipeptide that reversibly blocks the homodimerization of aminoimidazole carboxamide ribonucleotide transformylase/inosine nomophosphate cyclohydrolase (ATIC) and inhibits its activity (Ki = 691 nM, Kd = 240nM). AITC is a bifunctional enzyme that catalyzes the last two steps of de novo purine biosynthesis. Increases the intracellular levels of AICAR leading to the activation of AMP-activated protein kinase (AMPK). Shown to reduce cell viability in a dose-dependent manner. Exhibits a longer half-life and sustained AMPK activation (∽ 24 hours) compared to AICA-riboside (Cat. No. 123040).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins ATIC
Molecular Formula C21H33N7O5
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.