Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Inhibitor, Negative Control

Catalog No. 3420001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-200-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-200-01MG Supplier MilliporeSigma™ Supplier No. 3420001MG

missing translation for 'validMainframeMsg'

The Caspase Inhibitor, Negative Control, also referenced under CAS 105637-38-5, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

  • Peptide Sequence: Z-Phe-Ala-FMK
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Cathepsin-B
CAS 105637-38-5
Molecular Formula C21H23FN2O4
Formula Weight 386.4g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.