Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ATR Inhibitor IV

Catalog No. 5049720001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-497-20001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-497-20001 Supplier MilliporeSigma™ Supplier No. 5049720001
Add to Cart
Add to Cart

A cell-permeable phenylpyrazine derivative that acts as a potent, ATP-competitive ( Ki 13nM; IC50/[ATP] = 70 nM/50μM and 128 nM/100μM), ATR-selective inhibitor with much reduced or little potency against 4 related PIKKs

A cell-permeable phenylpyrazine derivative that acts as a potent, ATP-competitive ( K i= 13nM; IC50/[ATP] = 70 nM/50μM and 128 nM/100μM), ATR-selective inhibitor with much reduced or little potency against 4 related PIKKs ( K i= 2.2, 3.9, 16, and >1μM, respectively, against DNA-PK, PI 3-Kγ, ATM, and mTOR) and 50 other kinases. Shown to selectively inhibit ATR-dependent H2AX Ser139 and Chk1 Ser345 phosphorylation (complete inhibition at 10μM in HT29 and HFL1 cultures) without affecting ATM- and DNA-PK-mediated H2AX or ATM-mediated Chk2 phosphorylation. Short-term (24 h) VE-821 treatment, either alone or in synergy with Cisplatin (Cat. No. 232120), results in reversible cytostatic growth arrest regardless of cellular ATM-p53 pathway activity, while cytotoxicity and Cisplatin synergism in cell death induction is reported to occur only upon long-term VE-821 exposure (≥72 h) in cultures with ATM-p53 pathway defects.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins ATR
Molecular Formula C18H16N4O3S
Formula Weight 368.41g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.