Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ATR Kinase Inhibitor II

Catalog No. 11851010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-851-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-851-010MG Supplier MilliporeSigma™ Supplier No. 11851010MG
Add to Cart
Add to Cart

A cell-permeable aminopyrazinyl-sulfone compound that acts as a potent, reversible and ATP-competitive inhibitor of ATR kinase (IC50 = 12nM; Ki = 6nM) with excellent selectivity over ATM, DNA-PK (IC50 > 8μM) and in a 50-kinase panel (% inhibition at 2μM = 59, 57, 55, 52, 43 and 42 for AMPK, Lck, RIPK2, Ret, IKKα and FGFR3, resp

A cell-permeable aminopyrazinyl-sulfone compound that acts as a potent, reversible and ATP-competitive inhibitor of ATR kinase (IC50 = 12nM; Ki = 6nM) with excellent selectivity over ATM, DNA-PK (IC50 > 8μM) and in a 50-kinase panel (% inhibition at 2μM = 59, 57, 55, 52, 43 and 42 for AMPK, Lck, RIPK2, Ret, IKKα and FGFR3, respectively). Shown to block pH2AX-Ser139 (IC50 = 420nM in hydroxyurea treated HT29 cells) and preferentially enhance cisplatin (Cat. No. 232120) cytotoxicity against tumor cells over normal cells by ∽7-fold at 620 nM.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
Molecular Formula C22H25N5O3S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.