Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ATRA-BA Hybrid

Catalog No. 55472325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-472-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-472-325MG Supplier MilliporeSigma™ Supplier No. 55472325MG
Add to Cart
Add to Cart

The differentiation-inducer all-trans-Retinoic Acid and the histone deacetylase inhibitor (HDI) Butyric Acid.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Molecular Formula C31H40O4
Formula Weight 476.7g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.