Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Aurora Kinase/Cdk Inhibitor

Catalog No. 1894065MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-940-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-940-65MG Supplier MilliporeSigma™ Supplier No. 1894065MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Aurora Kinase/Cdk Inhibitor, also referenced under CAS 443797-96-4, controls the biological activity of Aurora Kinase/Cdk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins Aurora Kinase/Cdk
CAS 443797-96-4
Molecular Formula C15H12F2N6O3S·CH3CN
Formula Weight 435.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.