Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autocamtide-2 Related Inhibitory Peptide II, Cell-permeable

Catalog No. 1894851MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-948-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-948-51MG Supplier MilliporeSigma™ Supplier No. 1894851MG
Add to Cart
Add to Cart

The Autocamtide-2 Related Inhibitory Peptide II, Cell-permeable controls the biological activity of calmodulin-dependent protein kinase II. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Lys-Lys-Lys-Leu-Arg-Arg-Gln-Glu-Ala-Phe-Asp-Ala-Leu-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins cam Kinase-2
Molecular Formula C177H291N57O39S
Formula Weight 3873.7g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.