Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autocamtide-2 Related Inhibitory Peptide II

Catalog No. 1894841MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-948-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-948-41MG Supplier MilliporeSigma™ Supplier No. 1894841MG

The Autocamtide-2 Related Inhibitory Peptide II controls the biological activity of calmodulin-dependent protein kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Lys-Lys-Lys-Leu-Arg-Arg-Gln-Glu-Ala-Phe-Asp-Ala-Leu-OH
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins CAM Kinase-2
Molecular Formula C71H123N23O19
Formula Weight 1602.9g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.