Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autophagy Inducer, STF-62247

Catalog No. 18949710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-949-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-949-710MG Supplier MilliporeSigma™ Supplier No. 18949710MG
Add to Cart
Add to Cart

A cell-permeable autophagy inducer that selectively induces non-apoptotic cell death in VHL-deficient, but not VHL-expressing, renal carcinoma cells both in vitro and in mice in vivo.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 95% by HPLC
CAS 315702-99-9
Molecular Formula C15H13N3S
Formula Weight 267.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.