Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autophagy Inhibitor IV, Cpd18

Catalog No. 5059800001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-598-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-598-00001 Supplier MilliporeSigma™ Supplier No. 5059800001
Add to Cart
Add to Cart

A cell-permeable 3-methyladenine (3-MA) derivative that acts as an autophagy inhibitor via the inhibition of class III PI3K, and without touching class I PI3K

A cell-permeable 3-methyladenine (3-MA) derivative that acts as an autophagy inhibitor via the inhibition of class III PI3K, and without touching class I PI3K. Shown to block 80% of starvation-induced autophagy at 1mm (IC50 = 0.67 mM, compared with 3-MA at 6 mM). Displays much better solubility than 3-MA (5 times more soluble in water and 30 times more soluble in DMSO). Also found to reduce cell viability by 55% at 1mm at 12 hours compared with 72% reduction for 3-MA at 10 mM.

Please note that the molecular weight for this compound is lot-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins PI3K
Molecular Formula C12H17N5
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.