Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autotaxin Inhibitor I, S32826

Catalog No. 1895105MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-951-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-951-05MG Supplier MilliporeSigma™ Supplier No. 1895105MG
Add to Cart
Add to Cart

The Autotaxin Inhibitor I, S32826, also referenced under CAS 1103672-43-0, controls the biological activity of Autotaxin.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
CAS 1103672-43-0
Molecular Formula C21H34NO4PNa2·2H2O
Formula Weight 477.5g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.