Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autotaxin Inhibitor II, HA130

Catalog No. 18951110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-951-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-951-110MG Supplier MilliporeSigma™ Supplier No. 18951110MG
Add to Cart
Add to Cart

A thiazolidinedione that is designed to target the ATX active site thr210 with a boronic moiety and shown to potently inhibit ATX lysoPLD activity (IC50 ∽28nM using recombinant human ATX or human plasma) in a reversible manner with a mixed-type Lineweaver-Burk kinet

A thiazolidinedione that is designed to target the ATX active site thr210 with a boronic moiety and shown to potently inhibit ATX lysoPLD activity (IC50 ∽28nM using recombinant human ATX or human plasma) in a reversible manner with a mixed-type Lineweaver-Burk kinetics. Reported to exhibit low cytotoxicity (TD50 = 83, 105, and 2056μM in HEK293T, A2058, and HepG2 cultures) and no activity toward NPP1, PDE, AP, or proteasomal chymotryptic/tryptic/caspase-like activities even at concentrations as high as 10μM. Although HA130 quickly and effectively reduces circulating LPA level when injected in mice (463μg/kg, i.v.) in vivo , the effect appears short-lived as is the case with S32826 (Cat. No. 189510).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
Molecular Formula C24H19BFNO5S
Formula Weight 463.3g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.