Learn More
Description
- Cyclizes as it reduces disulfides to thiols, so reaction is "driven" to completion
- Does not contain waxy surfactant found in many other preparations
Specifications
Specifications
| Molecular Weight (g/mol) | 154.24 |
| Purity | ≥97% (Titration) |
| Molecular Formula | C4H10O2S2 |
| Content And Storage | 2°C to 8°C |
| Form | Powder |
| Quantity | 25 g |
| Synonym | DTT, Dithiothreitol |
| Solubility | EtOH, Water |
| Product Type | Calbiochem Cleland’s Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.