Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bax-Inhibiting Peptide, Negative Control

Catalog No. 1968115MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-681-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-681-15MG Supplier MilliporeSigma™ Supplier No. 1968115MG
Add to Cart
Add to Cart

A cell-permeable negative control of the Bax-Inhibiting Peptide, V5. Does not suppress Bax-mediated apoptosis (≈200 μM).

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Ile-Pro-Met-Ile-Lys-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Negative control of the Bax-Inhibiting Peptide, V5 (Cat. No. 196810)
Molecular Formula C28H52N6O6S
Formula Weight 600.8g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.