Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BAY 41-2272

Catalog No. 1968765MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-687-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-687-65MG Supplier MilliporeSigma™ Supplier No. 1968765MG
Add to Cart
Add to Cart

A cell-permeable pyrazolopyridinylpyrimidine compound that acts as a selective and potent stimulator of soluble guanylate cyclase (effective dose ∽ 0.1nM to 100μM using recombinant soluble guanylate cyclase).

A cell-permeable pyrazolopyridinylpyrimidine compound that acts as a selective and potent stimulator of soluble guanylate cyclase (effective dose ∽ 0.1nM to 100μM using recombinant soluble guanylate cyclase). The mode of activation is NO-independent and appears to be mediated through direct binding to the α1 and α2 subunits. Shown to be effective in treating numerous cardiovascular conditions in animal models. Inhibits phenylephrine-induced constriction of rabbit aortic rings (IC50 = 304nM) and blocks collagen-induced aggregation of human platelets (IC50 = 36nM). Does not inhibit phosphodiesterases.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Soluble guanylate cyclase
CAS 256376-24-6
Molecular Formula C20H17FN6
Formula Weight 360.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.