Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bcl-xL BH44-23, Human, Cell-Permeable

Catalog No. 1972171MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-721-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-721-71MG Supplier MilliporeSigma™ Supplier No. 1972171MG
Add to Cart
Add to Cart

A cell-permeable peptide that prevents apoptotic cell death by directly binding to the voltage-dependent anion channel (VDAC) and blocking its activity.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-βAla-Ser-Asn-Arg-Glu-Leu-Val-Val-Asp-Phe-Leu-Ser-Tyr-Lys-Ser-Gln-Lys-Gly-Tyr-Ser-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Voltage-dependent anion channel (VDAC)
Molecular Formula C165H279N59O46
Formula Weight 3825.4g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.