Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BEC, Hydrochloride

Catalog No. 1979005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-790-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-790-05MG Supplier MilliporeSigma™ Supplier No. 1979005MG
Add to Cart
Add to Cart

A boronic acid-based arginine analog that acts as a slow-binding, competitive transition state inhibitor of arginases I and II (Ki = 310 nM for human recombinant type II arginase, pH 7.5).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by NMR
Target Proteins arginase 1, arginase 2
Molecular Formula C5H12BNO4S·HCl
Formula Weight 229.5g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.