Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Benztropine

Catalog No. 5098900001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-989-00001 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-989-00001 Supplier MilliporeSigma™ Supplier No. 5098900001
Add to Cart
Add to Cart

An antagonist of muscarinic M1 and M3 receptors that induces robust differentiation of oligodendrocytes precursor cell differentiation (EC50 = 500nM) and promotes myelination.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Muscarinic M1/M3
Molecular Formula C21H2NO·CH3SO3H
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.