Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Berberine Chloride

Catalog No. 20027525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-027-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-027-525MG Supplier MilliporeSigma™ Supplier No. 20027525MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

A highly potent and selective oxysterol EBI2 (GPR183) agonist (K

A highly potent and selective oxysterol EBI2 (GPR183) agonist (Kd) = 450 pM in a saturation binding assay, and EC50 = 140 pM over EC50 = 2.1nM for its enantiomer, 7β,25-OHC, in a GTP-γS binding assay). Dose-dependently suppresses forskolin-induced cAMP accumulation in an EBI2-expressing SK-N-MC/CRE-β-galactosidase cell line (IC50 = 2nM), but not in control cells. Stimulates migration of LPS-activated spleen B-cells and anti-CD3/CD28-activated CD4+ T-cells in a dose-dependent manner. In addition, pharmacological inhibition of its biosynthesis in vivo by Clotrimazole, a CYP7B1inhibitor, promotes the migration of adoptively transferred pre-activated B cells to the T/B boundary, mimicking the phenotype of pre-activated B cells in EBI2-deficient mice.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC (NMR if no UV signal)
CAS 633-65-8
Molecular Formula C20H18NO4+Cl-
Formula Weight 371.81g/mol
Solubility DMSO or H2O
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.