Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (+)-Bicuculline

Catalog No. 5058750001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-587-50001 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-587-50001 Supplier MilliporeSigma™ Supplier No. 5058750001
Add to Cart
Add to Cart

A potent allosteric antagonist of vertebrate GABAA recept

A potent allosteric antagonist of vertebrate GABAA receptors. Blocks GABAA-induced chloride currents in cultured neurons (IC50 = 740nM). Shown to increase blood pressure in spontaneously hypertensive rats and cause convulsion in experimental animals. Also, increases the amplitude of spontaneous colonic contractions.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 99% by HPLC
Target Proteins GABAA
Molecular Formula C20H17NO6
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.