Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bisindolylmaleimide IV

Catalog No. 2032971MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-329-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-329-71MG Supplier MilliporeSigma™ Supplier No. 2032971MG
Add to Cart
Add to Cart

A potent, cell permeable, and selective inhibitor of protein kinase C (IC50 = 87 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 95% by HPLC
Target Proteins PKC
CAS 119139-23-0
Molecular Formula C20H13N3O2
Formula Weight 327.3g/mol
Solubility DMSO or MeOH
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.