Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (+)-Blebbistatin

Catalog No. 2033921MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-339-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-339-21MG Supplier MilliporeSigma™ Supplier No. 2033921MG
Add to Cart
Add to Cart

The inactive enantiomer of (±)-Blebbistatin.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by Chiral HPLC
Target Proteins Inactive enantiomer of (±)-Blebbistatin
CAS 1177356-70-5
Molecular Formula C18H16N2O2
Formula Weight 292.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.