Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BLM Helicase Inhibitor, ML216

Catalog No. 5063840001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-638-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-638-40001 Supplier MilliporeSigma™ Supplier No. 5063840001
Add to Cart
Add to Cart

A cell-permeable pyridinyl-thiadiazolylurea compound that selectively blocks Bloom's syndrome protein (BLM) helicase activity of both full-length as well as truncated BLM636-1298 forms (IC50

A cell-permeable pyridinyl-thiadiazolylurea compound that selectively blocks Bloom′s syndrome protein (BLM) helicase activity of both full-length as well as truncated BLM636-1298 forms (IC50 = 2.98 and 0.97μM, respectively). Competes with and reversibly blocks DNA binding to BLM. However, the inhibition appears to be non-competitive with respect to ATP. Exhibits anti-proliferative activity in cells expressing BLM, but not in those lacking BLM. Induces sister chromatid exchanges (SCEs) and enhances the sensitivity of BLM containing PSNF5 cells to aphidicolin (Cat. No. 178273) without affecting PSNG13 cells that are devoid of BLM. Displays favorable microsomal (T1/2 > 1.2 h) and plasma stability (∽100% at 5 h).
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 97% by HPLC
Target Proteins BLM
Molecular Formula C15H9F4N5OS
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.