Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BMP Inhibitor III, LDN-212854

Catalog No. 5054200001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-542-00001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-542-00001 Supplier MilliporeSigma™ Supplier No. 5054200001
Add to Cart
Add to Cart

A cell-permeable pyrazolopyrimidinylquinoline that is superior to Dorsomorphin (Cat

A cell-permeable pyrazolopyrimidinylquinoline that is superior to Dorsomorphin (Cat. Nos. 171260 and 171261) and DMH1 (Cat. No. 203646) as an ATP-site-targeting, ALK1/2 inhibitor (IC50 = 2.40 and 1.30 nM, respectively; [ATP] = 6μM) with respect to both potency and selectivity over ALK3/4/5 (IC50 = 0.858, 2.133, and 9.276μM, respectively). Also a potent RIPK2 inhibitor (IC50 = 6nM) with reduced potency against ABL1/c-abl, PDGFR-β, ALK3, and PDGFR-α (IC50 = 40, 68, 85.8, and 650 nM, respectively) and little inhibitory activity toward a panel of more than 190 kinases. Effectively inhibits ALK2- and ALK3-mediated C2C12 osteogenic differentiation (IC50 = 10 and 40.5 nM, respectively) in vitro and prevents constitutively active ALK2 Q207D mutant-induced heterotopic ossification in mice (6mg/kg/12 h via i.p.) in vivo. A good complement to the ALK4/5/7-selective A-83-01 (Cat. No. 616454).
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins ALK1/2/3
Molecular Formula C25H22N6
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.