Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BMX-IN-1

Catalog No. 5050210001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-502-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-502-10001 Supplier MilliporeSigma™ Supplier No. 5050210001
Add to Cart
Add to Cart

A cell-permeable tricyclic quinoline-acrylamide compound that potently inhibits BMX and BTK kinase activity (IC50 = 8 and

A cell-permeable tricyclic quinoline-acrylamide compound that potently inhibits BMX and BTK kinase activity (IC50 = 8 and 10.4 nM, respectively) by covalently targeting ATP-binding site cysteine, while being less potent against TEC family kinases TEC, JAK3, BLK with the same conserved cysteine (IC50 = 175, 377, and 653 nM, respectively). Only PI 3-Kγ/PIK3CG, SBK1, PIP5K2C (by ≥94% at 1μM), and TXK (by 75% at 1μM) are significantly inhibited by BMX-IN-1 in a selectivity profiling against 437 other kinase constructs. Potently inhibits the proliferation of TEL-BMX transformed Ba/F3 Ba/F3 (GI50 = 25nM), while being much less effective against TEL-BLK, TEL-JAK1, TEL-JAK2, TEL-JAK3, or TEL-TYK2E957D transformed Ba/F3 (GI50 = ≥3.64μM) or a panel of 5 prostate cancer cell lines (GI50 ≥2.46μM).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins BMX
Molecular Formula C29H24N4O4S
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.