Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bovine Plasma Fibronectin

Catalog No. 3416315MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-163-15MG 5 mg
34-163-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-163-15MG Supplier MilliporeSigma™ Supplier No. 3416315MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified from pooled bovine plasma.

EMD Millipore Calbiochem™ Bovine Plasma Fibronectin is an effective agent for promoting attachment of cells to commonly-used culture substrates.

  • Native form
  • Forms single major band in SDS-PAGE
  • CAS: 86088-83-7
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Bovine Plasma Fibronectin
Formulation In 150mM NaCl, 20mM sodium phosphate buffer, pH 7.3.
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.