Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bovine Serum Albumin, Fraction V (Fatty Acid, Nuclease and Protease Free)

Catalog No. 1266095GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-660-95GM 5 g
12-660-910GM 10 g
12-660-9100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-660-95GM Supplier MilliporeSigma™ Supplier No. 1266095GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use in serological testing, RIA, hormone response studies.

  • Contaminants
  • Ash: ≤3.0%
  • Fatty acids: ≤0.02%
  • Moisture: ≤5%
  • Nuclease: ≤0.1U/mg
  • Protease activity: ≤0.1U/mg
  • Heavy metals ≤10ppm
  • Assay: ≥98% by SDS-PAGE
  • CAS: 9048-46-8
  • Color: Beige

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.