Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Leupeptin, Hemisulfate, Synthetic

Catalog No. 10897610MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-897-610MG 10 mg
10-897-625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-897-610MG Supplier MilliporeSigma™ Supplier No. 10897610MG
Add to Cart
Add to Cart

Synthetic version of the microbial-derived inhibitor Leupeptin against trypsin-like and cysteine protease activities.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Leu-Leu-arginal, hemisulfate
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
CAS 103476-89-7
Molecular Formula C20H38N6O4·0.5H2SO4
Formula Weight 475.6g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Purity 95% by HPLC (sum of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.