Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ (+)-Brefeldin A, Eupenicillium brefeldianum
Shop All MilliporeSigma ProductsDescription
Specifically and reversibly blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus without affecting endocytosis or lysosome function. Inhibitor of HDL-mediated cholesterol efflux. Causes disassembly of the Golgi complex and ER swelling in a variety of mammalian cell lines at <40 ng/ml. Blocks binding of ADP-ribosylation factor to the Golgi and inhibits the GDP-GTP exchange. Inhibits the activity of BIG1 and BIG2, the guanine-nucleotide exchange proteins for ADP-ribosylation factors. Also available as a 25mm solution in DMSO (Cat. No. 500583).
Specifications
Specifications
| Color | White |
| Description | 95% by HPLC |
| Target Proteins | Blocks translocation of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus |
| CAS | 20350-15-6 |
| Molecular Formula | C16H24O4 |
| Formula Weight | 280.4g/mol |
| Solubility | MeOH |
| Form | Crystalline Solid |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?