Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Brevetoxin PbTx-3, Ptychodiscus brevis

Catalog No. 203734100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-373-4100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-373-4100UG Supplier MilliporeSigma™ Supplier No. 203734100UG
Add to Cart
Add to Cart

Lipid-soluble polyether marine toxin produced by the red tide dinoflagellate, Ptychodiscus brevis, found along the Gulf Coast of Florida.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Description >95% by HPLC
Target Proteins Voltage-dependent Na+ channel activator
CAS 85079-48-7
Molecular Formula C50H72O14
Formula Weight 897.1g/mol
Solubility MeOH or H2O
Form Solid
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.