Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bruton's Tyrosine Kinase Inhibitor III

Catalog No. 2036611MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-366-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-366-11MG Supplier MilliporeSigma™ Supplier No. 2036611MG

The Bruton's Tyrosine Kinase Inhibitor III, also referenced under CAS 936563-87-0, controls the biological activity of Bruton's Tyrosine Kinase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
CAS 936563-87-0
Molecular Formula C25H24N6O2
Formula Weight 440.5g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Crystalline Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.