Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bryostatin 1

Catalog No. 20381110UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-381-110UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-381-110UG Supplier MilliporeSigma™ Supplier No. 20381110UG
Add to Cart
Add to Cart

A macrocyclic lactone with antitumor properties.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description 98% by TLC
Target Proteins Protein kinase C
CAS 83314-01-6
Molecular Formula C47H68O17
Formula Weight 905.1g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid to Film
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.