Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BSA, Fraction V, Fatty Acid-Free

Catalog No. 12657510GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-657-510GM 10 g
12-657-5100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-657-510GM Supplier MilliporeSigma™ Supplier No. 12657510GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Useful for immunodiagnostic assays.

  • Fatty Acid free
  • pH: 6.8 to 7.4
  • Contaminants
  • Ash: ≤2.0%
  • Fatty acids: ≤0.05%
  • Moisture: ≤2%
  • Heavy metals ≤10ppm
  • Assay: ≥98% by Cellulose Acetate Electrophoresis
  • CAS: 9048-46-8
  • Color: Off-White

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.