Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ c-Raf Inhibitor

Catalog No. 47595820MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-595-820MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-595-820MG Supplier MilliporeSigma™ Supplier No. 47595820MG
Add to Cart
Add to Cart

The c-Raf Inhibitor controls the biological activity of c-Raf. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

A cell-permeable pyrazoloimidazole phenyl urea derivative that acts as a selective type II inhibitor of c-Raf. It shows an IC50 value of 50nM in an enzymatic assay, and comparable anti-proliferative activities to sorafenib against A375P (GI 50 = 2.68μM) and WM3629 (GI50 = 0.86μM) human melanoma cell lines. When screening against a panel of 30 different kinases at a concentration of 10μM, it exerts an inhibitory activity of 96% on c-Raf, but below 50% for most of the other kinases.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Molecular Formula C23H19F3N8O
Formula Weight 480.45g/mol
Solubility DMSO
Form Solid
Quantity 20 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.