Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CA-074

Catalog No. 2055301MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-553-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-553-01MG Supplier MilliporeSigma™ Supplier No. 2055301MG
Add to Cart
Add to Cart

CA-074, also referenced under CAS 134448-10-5, modulates the biological activity of cathepsin B. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

An epoxysuccinyl peptide that acts as a potent, irreversible, and specific inhibitor of cathepsin B (IC50 = 2.24nM for rat liver cathepsin B). Reduces ischemia-induced neuronal death. Exhibits lower membrane permeability when compared to CA-074 Me (Cat. No. 205531).
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless or White
Description 98% by HPLC
Target Proteins cathepsin B
Molecular Formula C18H29N3O6
Formula Weight 383.4g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.