Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calcium Entry Blocker Set, Pyr6 & Pyr10

Catalog No. 5097090001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-970-90001 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-970-90001 Supplier MilliporeSigma™ Supplier No. 5097090001
Add to Cart
Add to Cart

A set of PYR6, a CRAC Channel Inhibitor III; and PYR10, a TPRC3 Channel Inhibitor III.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A set consists of 10mg each of CRAC Channel Inhibitor III, PYR6 and TPRC3 Channel Inhibitor III, PYR10
Description 96% by HPLC for each compound
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.