Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calmodulin Kinase IINtide, Myristoylated

Catalog No. 2089211MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-892-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-892-11MG Supplier MilliporeSigma™ Supplier No. 2089211MG
Add to Cart
Add to Cart

The myristoylated form of CaMK IINtide.

  • Packaged Under Inert Gas
  • Peptide Sequence: Myr-N-Gly-Gly-Gly-Lys-Arg-Pro-Pro-Lys-Leu-Gly-Gln-Ile-Gly-Arg-Ala-Lys-Arg-Val-Val-Ile-Glu-Asp-Asp-Arg-Ile-Asp-Asp-Val-Leu-Lys-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Calmodulin-Dependent Protein Kinase (CaM Kinase)-2
Molecular Formula C256H275N47O43
Formula Weight 4698.3g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.