Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calmodulin Kinase IINtide

Catalog No. 2089201MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-892-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-892-01MG Supplier MilliporeSigma™ Supplier No. 2089201MG
Add to Cart
Add to Cart

A potent and specific inhibitor of CaMK II (IC50 = 50 nM for both the total and Ca2+-independent CaMK II activity).

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Lys-Arg-Pro-Pro-Lys-Leu-Gly-Gln-Ile-Gly-Arg-Ala-Lys-Arg-Val-Val-Ile-Glu-Asp-Asp-Arg-Ile-Asp-Asp-Val-Leu-Lys-OH
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Cam Kinase-2
Molecular Formula C136H240N44O39
Formula Weight 3115.7g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.