Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpain Inhibitor IV

Catalog No. 2087241MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-872-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-872-41MG Supplier MilliporeSigma™ Supplier No. 2087241MG
Add to Cart
Add to Cart

The Calpain Inhibitor IV controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Peptide Sequence: Z-Leu-Leu-Tyr-CH2F
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White to Pale Yellow
Description single spot by TLC
Target Proteins calpain 2
Molecular Formula C30H40FN3O6
Formula Weight 557.7g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.