Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpain Substrate II, Fluorogenic

Catalog No. 20873125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-873-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-873-125MG Supplier MilliporeSigma™ Supplier No. 20873125MG
Add to Cart
Add to Cart

Cyclic fluorogenic substrate for the quantitative determination of calpain -1 and -2.

Peptide Sequence: Suc-Leu-Tyr-AMC
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by TLC
Target Proteins Calpain -1 and -2
Molecular Formula C29H31N3O7
Formula Weight 533.6g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.