Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpain Substrate III, Fluorogenic

Catalog No. 2087711MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-877-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-877-11MG Supplier MilliporeSigma™ Supplier No. 2087711MG
Add to Cart
Add to Cart

A fluorogenic FRET peptide with a substrate sequence that is optimized for calpain-1 and -2.

  • Packaged Under Inert Gas
  • Peptide Sequence: DABCYL-Thr-Pro-Leu-Lys∼Ser-Pro-Pro-Pro-Ser-Pro-Arg-EDANS
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Formulation Supplied as a trifluoroacetate salt.
Description 98% by HPLC
Target Proteins Substrate sequence that is optimized for calpain-1 and -2
Molecular Formula C80H114N20O18S
Formula Weight 1676g/mol
Solubility DMSO or H2O
Content And Storage Protect from light
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.