Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpastatin, Human, Recombinant, Domain I

Catalog No. 2089001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-890-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-890-01MG Supplier MilliporeSigma™ Supplier No. 2089001MG
Only null left
Add to Cart
Add to Cart

Domain I of human calpastatin.

Domain I of human calpastatin. Endogenous protease inhibitor that acts specifically on Calpain-1 (Cat. No. 208712). Exhibits greater inhibitory potency than calpain inhibitors I and II.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description homogeneous by SDS-PAGE
Target Proteins calpain-1
Formula Weight 14000g/mol
Solubility H2O and/or Aqueous buffers
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.