Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpastatin Peptide, Negative Control

Catalog No. 208904500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-890-4500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-890-4500UG Supplier MilliporeSigma™ Supplier No. 208904500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A scrambled peptide with an identical amino acid composition to that of Calpastatin Peptide. Useful as a negative control for Calpastatin Peptide.

Peptide Sequence: Ac-Ala-Pro-Arg-Leu-Glu-Ile-Val-Pro-Thr-Met-Tyr-Ile-Tyr-Lys-Leu-Ser-Pro-Thr-Gly-Ser-Glu-Lys-Leu-Glu-Asp-Glu-Arg-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Useful as a negative control for Calpastatin Peptide
Molecular Formula C142H230N36O44S
Formula Weight 3177.7g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.