Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpeptin

Catalog No. 033400515MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-340-0515MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-340-0515MG Supplier MilliporeSigma™ Supplier No. 033400515MG

missing translation for 'validMainframeMsg'

A cell-permeable calpain inhibitor (ID50 = 52 nM for calpain-1; ID50 = 34 nM for calpain-2; ID50 = 138 nM for papain).

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Leu-Nle-CHO
  • Cell Permeable
  • Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Target Proteins Calpain-1
CAS 117591-20-5
Molecular Formula C20H30N2O4
Formula Weight 362.5g/mol
Solubility DMSO or DMF
Content And Storage Store at -20°C
Form Solid
Quantity 5 mg
Purity 94% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.