Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bradykinin

Catalog No. 052305001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-230-5001MG 1 mg
05-230-50025M 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-230-5001MG Supplier MilliporeSigma™ Supplier No. 052305001MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Induces the release of nitric oxide.

Peptide Sequence: H-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Formulation Supplied as an acetate salt.
Target Proteins Induces the release of nitric oxide
CAS 58-82-2
Molecular Formula C50H73N15O11
Formula Weight 1060.2g/mol
Solubility 5% Acetic Acid
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Purity 97% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.