Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Substance P

Catalog No. 0523060025M Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-230-60025M 25 mg
05-230-6001MG 1 mg
05-230-6005MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-230-60025M Supplier MilliporeSigma™ Supplier No. 0523060025MG
Add to Cart
Add to Cart

Neuropeptide found in high concentrations in the gut.

Peptide Sequence: H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Target Proteins Substance P receptors
CAS 33507-63-0
Molecular Formula C63H98N18O13S
Formula Weight 1347.6g/mol
Solubility 5% Acetic Acid
Content And Storage Hygroscopic
Form Solid
Quantity 25 mg
Purity 96% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.