Learn More
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | Clear to White |
| Description | 98% by HPLC |
| Product Type | Phosphatase Inhibitor |
| Target Proteins | PP2A |
| CAS | 101932-71-2 |
| Molecular Formula | C50H81N4O15P |
| Formula Weight | 1009.2g/mol |
| Solubility | DMSO or EtOH |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Film to Crystalline Powder |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.