Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CaMKII Inhibitor, CK59

Catalog No. 2089225MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-892-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-892-25MG Supplier MilliporeSigma™ Supplier No. 2089225MG
Add to Cart
Add to Cart

The CaMKII Inhibitor, CK59, also referenced under CAS 140651-18-9, controls the biological activity of CaMKII. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
CAS 140651-18-9
Molecular Formula C21H37N7O3
Formula Weight 435.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Semi-solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.