Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CaMKP Inhibitor

Catalog No. 208775100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-877-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-877-5100MG Supplier MilliporeSigma™ Supplier No. 208775100MG

missing translation for 'validMainframeMsg'

The CaMKP Inhibitor, also referenced under CAS 52789-62-5, controls the biological activity of CaMKP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 85% by Titration
CAS 52789-62-5
Molecular Formula C10H8NNaO7S2·2H2O
Formula Weight 377.3g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.